accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12401-56-8

IUPAC Name: hafnium disilicon
Synonyms: hafnium disilicon
Molecular Weight (g/mol): 234.66
Molecular Formula: HfSi2
InChi Key: TWRSDLOICOIGRH-UHFFFAOYSA-N
SMILES: [Si].[Si].[Hf]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results