accessibility menu, dialog, popup

UserName

Viewing profile as user:
SFFFFFFNH2

CAS RN 1240257-25-3

IUPAC Name: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 237.16
Molecular Formula: C6H5F6NS
InChi Key: VNFMUDAEGKTTOA-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results