accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1242258-38-3

IUPAC Name: 1-bromo-4-(difluoromethoxy)-2-fluorobenzene
Synonyms: 1-bromo-4-(difluoromethoxy)-2-fluorobenzene
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: QWUYTYZPBMAYKP-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results