accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHO

CAS RN 124344-98-5

IUPAC Name: (1-methyl-5-phenyl-1H-pyrazol-3-yl)methanol
Synonyms: (1-methyl-5-phenylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: ZMDMCKKOZJKHKG-UHFFFAOYSA-N
SMILES: CN1N=C(CO)C=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results