CAS RN 124432-70-8
CAS RN 124432-70-8
IUPAC Name:
5-bromo-3-fluoro-2-methoxypyridine
Synonyms:
5-bromo-3-fluoro-2-methoxypyridine
Molecular Weight (g/mol):
206.01
Molecular Formula:
C6H5BrFNO
InChi Key:
DEBFRAAJISQBIE-UHFFFAOYSA-N
SMILES:
COC1=NC=C(Br)C=C1F
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-Bromo-3-fluoro-2-methoxypyridine, 97%, Thermo Scientific™
CAS: 124432-70-8 Molecular Formula: C6H5BrFNO Molecular Weight (g/mol): 206.014 MDL Number: MFCD04112553 InChI Key: DEBFRAAJISQBIE-UHFFFAOYSA-N PubChem CID: 14837011 IUPAC Name: 5-bromo-3-fluoro-2-methoxypyridine SMILES: COC1=NC=C(C=C1F)Br