accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHO

CAS RN 1244954-13-9

IUPAC Name: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6-triene-1-carboxylic acid
Synonyms: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6-triene-1-carboxylic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: ZRBQIRRMWRKZTO-UHFFFAOYNA-N
SMILES: OC(=O)C12CCC(O1)C1=CC=CC=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results