CAS RN 1245-13-2
IUPAC Name:
[2,2'-biquinoline]-4,4'-dicarboxylic acid
Synonyms:
bicinchoninic acid
Molecular Weight (g/mol):
344.33
Molecular Formula:
C20H12N2O4
InChi Key:
AFYNADDZULBEJA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C(O)=O)=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2,2'-Bicinchoninic Acid 98.0+%, TCI America™
CAS: 1245-13-2 Molecular Formula: C20H12N2O4 Molecular Weight (g/mol): 344.326 MDL Number: MFCD00068342 InChI Key: AFYNADDZULBEJA-UHFFFAOYSA-N PubChem CID: 71068 IUPAC Name: 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid SMILES: C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O