accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOFFF

CAS RN 1245648-32-1

IUPAC Name: tert-butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 267.25
Molecular Formula: C11H16F3NO3
InChi Key: RSFRBHIRISNXIK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(=O)CC1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results