accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124655-09-0

IUPAC Name: tert-butyl 2-[6-(hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Synonyms: tert-butyl 2-[6-(hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molecular Weight (g/mol): 260.33
Molecular Formula: C13H24O5
InChi Key: CFRUAOXMCVQMFP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC1CC(CO)OC(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results