accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C(R)OHONHOOCH3CH3H3C

CAS RN 124655-17-0

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: LRFZIPCTFBPFLX-JLDDOWRYNA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](C(O)=O)C(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)