accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3N

CAS RN 1246669-45-3

IUPAC Name: 9-phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 369.27
Molecular Formula: C24H24BNO2
InChi Key: WUXIRZBTTICLCG-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results