CAS RN 124687-44-1
CAS RN 124687-44-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Bismuth(III) tert-pentyloxide, 97+%
CAS: 124687-44-1 MDL Number: MFCD00151666 Synonym: Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide