accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124733-24-0

IUPAC Name: 1,1,2,2-tetramethyl-1,2-bis(thiophen-2-yl)disilane
Synonyms: 1,1,2,2-tetramethyl-1,2-bis(thiophen-2-yl)disilane
Molecular Weight (g/mol): 282.57
Molecular Formula: C12H18S2Si2
InChi Key: FGYSQQKHYAVZEZ-UHFFFAOYSA-N
SMILES: C[Si](C)(C1=CC=CS1)[Si](C)(C)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results