accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONOHOHOHHOHCl

CAS RN 124763-51-5

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol hydrochloride
Synonyms: bis-tris buffer hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C8H20ClNO5
InChi Key: VEYRVLHAMHQVTC-UHFFFAOYSA-N
SMILES: Cl.OCCN(CCO)C(CO)(CO)CO
Grade 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)