accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1248656-98-5

IUPAC Name: λ²-iron(2+) 1-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide chloropalladiumylium pentaphenylcyclopenta-2,4-dien-1-ide prop-2-en-1-ide
Synonyms: λ²-iron(2+) 1-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide chloropalladiumylium pentaphenylcyclopenta-2,4-dien-1-ide prop-2-en-1-ide
Molecular Weight (g/mol): 893.67
Molecular Formula: C51H52ClFePPd
InChi Key: SURRVSCVVZPMFV-UHFFFAOYSA-M
SMILES: [Fe++].Cl[Pd+].[CH2-]C=C.CC(C)(C)P([C-]1C=CC=C1)C(C)(C)C.C1=CC=C(C=C1)[C-]1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results