accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CFFFFF

CAS RN 124898-13-1

IUPAC Name: trimethyl(1,1,2,2,2-pentafluoroethyl)silane
Synonyms: trimethyl(1,1,2,2,2-pentafluoroethyl)silane
Molecular Weight (g/mol): 192.2
Molecular Formula: C5H9F5Si
InChi Key: MTPVUVINMAGMJL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)