CAS RN 1249-97-4
IUPAC Name:
10-benzoyl-N3,N3,N7,N7-tetramethyl-10H-phenothiazine-3,7-diamine
Synonyms:
10-benzoyl-N3,N3,N7,N7-tetramethylphenothiazine-3,7-diamine
Molecular Weight (g/mol):
389.52
Molecular Formula:
C23H23N3OS
InChi Key:
ZKURGBYDCVNWKH-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C2N(C(=O)C3=CC=CC=C3)C3=CC=C(C=C3SC2=C1)N(C)C
1
–
1
of
1
results
1
Benzoyl Leuco Methylene Blue 96.0+%, TCI America™
CAS: 1249-97-4 Molecular Formula: C23H23N3OS Molecular Weight (g/mol): 389.517 MDL Number: MFCD00059153 InChI Key: ZKURGBYDCVNWKH-UHFFFAOYSA-N PubChem CID: 94975 IUPAC Name: [3,7-bis(dimethylamino)phenothiazin-10-yl]-phenylmethanone SMILES: CN(C)C1=CC2=C(C=C1)N(C3=C(S2)C=C(C=C3)N(C)C)C(=O)C4=CC=CC=C4