accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOONCH3OH3COH3C

CAS RN 124931-12-0

IUPAC Name: diethyl {[methoxy(methyl)carbamoyl]methyl}phosphonate
Synonyms: diethyl [methoxy(methyl)carbamoyl]methylphosphonate
Molecular Weight (g/mol): 239.21
Molecular Formula: C8H18NO5P
InChi Key: WYLRYBDGIILFIR-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC(=O)N(C)OC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results