accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3CFONH

CAS RN 1256255-84-1

IUPAC Name: 7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 279.12
Molecular Formula: C14H19BFNO3
InChi Key: OISLTYSRSUFGPW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results