CAS RN 125662-60-4
IUPAC Name:
2,3,5,6-tetrafluoro-4-sulfobenzoic acid
Synonyms:
2,3,5,6-tetrafluoro-4-sulfobenzoic acid
Molecular Weight (g/mol):
274.14
Molecular Formula:
C7H2F4O5S
InChi Key:
JNESNWCWZVATQY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(F)=C(C(F)=C1F)S(O)(=O)=O
1
–
1
of
1
results
1
4-Sulfo-2,3,5,6-tetrafluorobenzoic Acid 95.0+%, TCI America™
CAS: 125662-60-4 Molecular Formula: C7H2F4O5S Molecular Weight (g/mol): 274.142 InChI Key: JNESNWCWZVATQY-UHFFFAOYSA-N PubChem CID: 19807654 IUPAC Name: 2,3,5,6-tetrafluoro-4-sulfobenzoic acid SMILES: C1(=C(C(=C(C(=C1F)F)S(=O)(=O)O)F)F)C(=O)O