accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 125697-91-8

IUPAC Name: 5-{bis[(2-hydroxyphenyl)methyl]amino}-2-hydroxybenzoic acid
Synonyms: 5-{bis[(2-hydroxyphenyl)methyl]amino}-2-hydroxybenzoic acid
Molecular Weight (g/mol): 365.39
Molecular Formula: C21H19NO5
InChi Key: RTYOLBQXFXYMKY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1O)N(CC1=CC=CC=C1O)CC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results