accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CONONCH3CH3FBFFF

CAS RN 125700-71-2

IUPAC Name: [(dimethylamino)[(2-oxo-1,2-dihydropyridin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms: [(dimethylamino)[(2-oxopyridin-1-yl)oxy]methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 297.06
Molecular Formula: C10H16BF4N3O2
InChi Key: CZQGINAUZYECAI-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1C=CC=CC1=O)=[N+](C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results