accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1257251-75-4

IUPAC Name: 9-(3-bromophenyl)-9-phenyl-9H-fluorene
Synonyms: 9-(3-bromophenyl)-9-phenylfluorene
Molecular Weight (g/mol): 397.32
Molecular Formula: C25H17Br
InChi Key: LWBYEDXZPBXTFL-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results