accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 125867-25-6

IUPAC Name: 2-(2,2-dimethylpropanamido)pyridine-3-carboxylate
Synonyms: 2-(2,2-dimethylpropanamido)pyridine-3-carboxylate
Molecular Weight (g/mol): 221.24
Molecular Formula: C11H13N2O3
InChi Key: KXLOGMIKCNUSNG-UHFFFAOYSA-M
SMILES: CC(C)(C)C(=O)NC1=NC=CC=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results