accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFPFFFFFFFFFFFF

CAS RN 1259-35-4

IUPAC Name: tris(2,3,4,5,6-pentafluorophenyl)phosphane
Synonyms: tris(2,3,4,5,6-pentafluorophenyl)phosphane
Molecular Weight (g/mol): 532.15
Molecular Formula: C18F15P
InChi Key: FQLSDFNKTNBQLC-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)