accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1259280-37-9

IUPAC Name: 2-(9,9-diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(9,9-diphenylfluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 444.38
Molecular Formula: C31H29BO2
InChi Key: WTFLXHPXCVWFEP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C3=CC=CC=C3C(C2=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results