accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126-39-6

IUPAC Name: 2-ethyl-2-methyl-1,3-dioxolane
Synonyms: 2-ethyl-2-methyl-1,3-dioxolane
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: UPZFLZYXYGBAPL-UHFFFAOYSA-N
SMILES: CCC1(C)OCCO1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)