accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126-75-0

IUPAC Name: diethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonate
Synonyms: demeton demeton-S
Molecular Weight (g/mol): 258.33
Molecular Formula: C8H19O3PS2
InChi Key: GRPRVIYRYGLIJU-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)SCCSCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results