accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH2N

CAS RN 126-98-7

IUPAC Name: 2-methylprop-2-enenitrile
Synonyms: methacrylonitrile
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: GYCMBHHDWRMZGG-UHFFFAOYSA-N
SMILES: CC(=C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results