accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONBrN

CAS RN 1260762-06-8

IUPAC Name: 4-bromo-2-(morpholin-4-yl)benzonitrile
Synonyms: 4-bromo-2-(morpholin-4-yl)benzonitrile
Molecular Weight (g/mol): 267.13
Molecular Formula: C11H11BrN2O
InChi Key: SEDCSLWVDKMJCF-UHFFFAOYSA-N
SMILES: BrC1=CC(N2CCOCC2)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results