accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOCl

CAS RN 1260897-00-4

IUPAC Name: 1-(2-chloro-4-nitrophenyl)azepane
Synonyms: 1-(2-chloro-4-nitrophenyl)azepane
Molecular Weight (g/mol): 254.71
Molecular Formula: C12H15ClN2O2
InChi Key: YQJCKUFYSNESGZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(C=C1)N1CCCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results