CAS RN 126162-38-7
IUPAC Name:
2,3-difluoro-4-hydroxybenzonitrile
Synonyms:
2,3-difluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol):
155.1
Molecular Formula:
C7H3F2NO
InChi Key:
UIJJJWADIVZXNT-UHFFFAOYSA-N
SMILES:
OC1=C(F)C(F)=C(C=C1)C#N
1
–
1
of
1
results
1
Promotions Available
2,3-Difluoro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals
2,3-Difluoro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals
CAS: 126162-38-7 Molecular Formula: C7H3F2NO Molecular Weight (g/mol): 155.10 MDL Number: MFCD00269596 InChI Key: UIJJJWADIVZXNT-UHFFFAOYSA-N PubChem CID: 2736978 IUPAC Name: 2,3-difluoro-4-hydroxybenzonitrile SMILES: OC1=C(F)C(F)=C(C=C1)C#N