accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1261673-70-4

IUPAC Name: 2-chloro-6-(trifluoromethoxy)aniline
Synonyms: 2-chloro-6-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 211.57
Molecular Formula: C7H5ClF3NO
InChi Key: NNDFBODLWFJOSA-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)