accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12619-68-0

IUPAC Name: tris(λ²-cobalt(2+)) diboron
Synonyms: tris(λ²-cobalt(2+)) diboron
Molecular Weight (g/mol): 198.42
Molecular Formula: B2Co3
InChi Key: QSOMCIJHXYGMAO-UHFFFAOYSA-N
SMILES: [B].[B].[Co++].[Co++].[Co++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results