CAS RN 1262198-07-1
IUPAC Name:
3-bromo-2,6-difluoroaniline
Synonyms:
3-bromo-2,6-difluoroaniline
Molecular Weight (g/mol):
208.01
Molecular Formula:
C6H4BrF2N
InChi Key:
HIZADWSSPWORHH-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=CC(Br)=C1F
1
–
1
of
1
results
1
Promotions Available
3-Bromo-2,6-difluoroaniline, 96%
3-Bromo-2,6-difluoroaniline, 96%
CAS: 1262198-07-1 Molecular Formula: C6H4BrF2N Molecular Weight (g/mol): 208.01 MDL Number: MFCD22627833 InChI Key: HIZADWSSPWORHH-UHFFFAOYSA-N Synonym: benzenamine,3-bromo-2,6-difluoro PubChem CID: 21645692 IUPAC Name: 3-bromo-2,6-difluoroaniline SMILES: NC1=C(F)C=CC(Br)=C1F