accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOOONNH(S)(R)H3CH3CCH3

CAS RN 1262308-49-5

IUPAC Name: (4S,5R)-3-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S,5R)-3-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetyl)-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 438.48
Molecular Formula: C24H26N2O6
InChi Key: NUXZZZZUNKUMGM-SZNDQCEHSA-N
SMILES: C[C@H]1OC(C)(C)N([C@@H]1C(O)=O)C(=O)CNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)