accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNBrHBr

CAS RN 1262769-75-4

IUPAC Name: 3-bromopyrrolidine hydrobromide
Synonyms: 3-bromopyrrolidine hydrobromide
Molecular Weight (g/mol): 230.93
Molecular Formula: C4H9Br2N
InChi Key: ZSDJSWROXIUGAE-UHFFFAOYNA-N
SMILES: Br.BrC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results