CAS RN 1262776-24-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PESTANAL™ Clothianidin-d3, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Find more information here: Neonicotinoids