accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1263045-16-4

IUPAC Name: 1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid
Synonyms: 1-[3-(2,5-dioxopyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid
Molecular Weight (g/mol): 416.43
Molecular Formula: C18H28N2O9
InChi Key: IAJVEYLYYHOZEY-UHFFFAOYSA-N
SMILES: OC(=O)CCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results