accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126325-46-0

IUPAC Name: 2-amino-3-bromo-6-methylpyridin-1-ium
Synonyms: 2-amino-3-bromo-6-methylpyridin-1-ium
Molecular Weight (g/mol): 188.05
Molecular Formula: C6H8BrN2
InChi Key: JYWWGZAAXTYNRN-UHFFFAOYSA-O
SMILES: CC1=CC=C(Br)C(N)=[NH+]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results