accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HCHCZr

CAS RN 12636-72-5

IUPAC Name: zirconium(4+) bis(cyclopenta-2,4-dien-1-ide) dimethanide
Synonyms: zirconium(4+) bis(cyclopentadienide) dimethanide
Molecular Weight (g/mol): 251.48
Molecular Formula: C12H16Zr
InChi Key: FOJOHGYNJWTFHE-UHFFFAOYSA-N
SMILES: [CH3-].[CH3-].[Zr+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results