accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126370-67-0

IUPAC Name: 6-phenylpyridin-3-amine
Synonyms: 6-phenylpyridin-3-amine
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: FLIQYTXJLWGVBG-UHFFFAOYSA-N
SMILES: NC1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results