accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12645-31-7

H3CCH3OHOHPOHOHO

CAS RN 12645-31-7

IUPAC Name: 2-ethylhexan-1-ol; phosphoric acid
Synonyms: 2-ethylhexanol; phosphoric acid
Molecular Weight (g/mol): 228.23
Molecular Formula: C8H21O5P
InChi Key: FZQYZMODYSBPES-UHFFFAOYNA-N
SMILES: OP(O)(O)=O.CCCCC(CC)CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)