accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(S)(R)HO

CAS RN 126456-43-7

IUPAC Name: (1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Synonyms: (1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Molecular Weight (g/mol): 150.2
Molecular Formula: C9H12NO
InChi Key: LOPKSXMQWBYUOI-BDAKNGLRSA-O
SMILES: [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (7)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)