accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1265140-81-5

IUPAC Name: dichloropalladium; dicyclohexyl({4-[4-(dicyclohexylphosphanyl)phenoxy]phenyl})phosphane
Synonyms: dicyclohexyl({4-[4-(dicyclohexylphosphanyl)phenoxy]phenyl})phosphane; palladium chloride
Molecular Weight (g/mol): 740.08
Molecular Formula: C36H52Cl2OP2Pd
InChi Key: UPHBRQDWOFHFER-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1=CC=C(OC2=CC=C(C=C2)P(C2CCCCC2)C2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results