accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSNNO

CAS RN 126533-95-7

IUPAC Name: ethyl 2-(morpholin-4-yl)-1,3-thiazole-4-carboxylate
Synonyms: ethyl 2-(morpholin-4-yl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 242.29
Molecular Formula: C10H14N2O3S
InChi Key: OINWWRRVBHJAKO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results