accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1266350-99-5

OOOOHOOONNH(S)(R)H3CH3CCH3(S)H3CH3CCH3

CAS RN 1266350-99-5

IUPAC Name: (4S,5R)-3-[(2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S,5R)-3-[(2S)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 524.61
Molecular Formula: C29H36N2O7
InChi Key: AVAILEBHPYBEGO-CQLNOVPUSA-N
SMILES: C[C@H]1OC(C)(C)N([C@@H]1C(O)=O)C(=O)[C@H](COC(C)(C)C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)