accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOFFNNNFF

CAS RN 126695-58-7

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-azido-2,3,5,6-tetrafluorobenzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 4-azido-2,3,5,6-tetrafluorobenzoate
Molecular Weight (g/mol): 332.17
Molecular Formula: C11H4F4N4O4
InChi Key: HCTLWQSYGIBOFM-UHFFFAOYSA-N
SMILES: FC1=C(F)C(C(=O)ON2C(=O)CCC2=O)=C(F)C(F)=C1N=[N+]=[N-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results