accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12672-27-4

IUPAC Name: dialuminium(3+) bis(λ²-cobalt(2+)) pentaoxidandiide
Synonyms: dialuminium(3+) bis(λ²-cobalt(2+)) pentaoxidandiide
Molecular Weight (g/mol): 251.82
Molecular Formula: Al2Co2O5
InChi Key: CRHLEZORXKQUEI-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Co++].[Co++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results