accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClCl

CAS RN 12672-29-6

IUPAC Name: 3,3',5,5'-tetrachloro-1,1'-biphenyl
Synonyms: 3,3',5,5'-tetrachlorobiphenyl 3,3',5,5'-tetrachlorodiphenyl
Molecular Weight (g/mol): 291.98
Molecular Formula: C12H6Cl4
InChi Key: UTMWFJSRHLYRPY-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)C1=CC(Cl)=CC(Cl)=C1
Solvent or Matrix 
  • (1)
  • (1)
Technique 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results